Learn More
Invitrogen™ Nucleostemin Monoclonal Antibody (2C8D5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517082
Description
MA5-17082 targets GNL3 in FACS, indirect ELISA, and WB applications and shows reactivity with Human and Mouse samples. The MA5-17082 immunogen is purified recombinant fragment of human GNL3 (amino acids: 1-226) expressed in E. Coli. MA5-17082 detects GNL3 which has a predicted molecular weight of approximately 62kDa.
Arylsulfatase F, also known as ARSF, is a 590 amino acid secretory protein that belongs to the sulfatase family of bone and cartilage matrix proteins. Arylsulfatase F uses calcium as a cofactor to catalyze reactions that are important in maintaining correct bone composition. The activity of Arylsulfatase F, unlike that of other family members, such as Arylsulfatase E, is not inhibited by warfarin. The gene encoding Arylsulfatase F maps to human chromosome X, which contains nearly 153 million base pairs and houses over 1,000 genes. In conjunction with chromosome Y, chromosome X is responsible for sex determination. There are a number of conditions related to an abnormal number and combination of sex chromosomes, some of which include Turner's syndrome, color blindness, hemophilia and Duchenne muscular dystrophy.
Specifications
| Nucleostemin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| Q8CI11, Q9BVP2 | |
| GNL3 | |
| Purified recombinant fragment of human GNL3 (amino acids: 1-226) expressed in E. Coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 2C8D5 | |
| Unconjugated | |
| GNL3 | |
| C77032; E2IG3; E2-induced gene 3 protein; estradiol-induced nucleotide binding protein; G protein nucleolar 3; Gnl3; guanine nucleotide binding protein-like 3 (nucleolar); guanine nucleotide-binding protein-like 3; MGC800; NNP47; novel nucleolar protein 47; Ns; nucleolar GTP-binding protein 3; Nucleostemin | |
| Mouse | |
| Protein G | |
| RUO | |
| 26354, 30877 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.