Learn More
GDP-mannose pyrophosphorylase A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human GMPPA protein.
Brand: Abnova H00029926-D01P.100ug
Description
This gene is thought to encode a GDP-mannose pyrophosphorylase. This enzyme catalyzes the reaction which converts mannose-1-phosphate and GTP to GDP-mannose which is involved in the production of N-linked oligosaccharides. [provided by RefSeq
Sequence: MLKAVILIGGPQKGTRFRPLSFEVPKPLFPVAGVPMIQHHIEACAQVPGMQEILLIGFYQPDEPLTQFLEAAQQEFNLPVRYLQEFAPLGTGGGLYHFRDQILAGSPEAFFVLNADVCSDFPLSAMLEAHRRQRHPFLLLGTTANRTQSLNYGCIVENPQTHEVLHYVEKPSTFISDIINCGIYLFSPEALKPLRDVFQRNQQDGQLEDSPGLWPGAGTIRLEQDVFSALAGQGQIYVHLTDGIWSQIKSAGSALYASRLYLSRYQDTHPERLAKHTPGGPWIRGNVYIHPTAKVAPSAVLGPNVSIGKGVTVGEGVRLRESIVLHGATLQEHTCVLHSIVGWGSTVGRWARVEGTPSDPNPNDPRARMDSESLFKDGKLLPAITILGCRVRIPAEVLILNSIVLPHKELSRSFTNQIILSpecifications
| GDP-mannose pyrophosphorylase A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human GMPPA protein. | |
| GMPPA | |
| GMPPA | |
| GMPPA (NP_037467.2, 1 a.a. ∼ 420 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_013335.2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 29926 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.