Learn More
GM130 Mouse, Unlabeled, Clone: 35/GM130, BD
Mouse Monoclonal Antibody
£292.00 - £499.00
Specifications
| Antigen | GM130 |
|---|---|
| Clone | 35/GM130 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15855659
|
BD Biosciences
610822 |
50 μg |
£292.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15865659
|
BD Biosciences
610823 |
150 μg |
£499.00
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Maturation and post-translational modification of proteins occurs after their biosynthesis at the endoplasmic reticulum and their transport through the Golgi apparatus. The process involves the transport of vesicles carrying the proteins through a vectorial process of vesicle budding and fusion from the cis-compartment to the medial-compartment and the trans-compartment of the Golgi apparatus. The detergent insoluble fraction of the Golgi is named “matrix” and is required for proper morphology of the Golgi membranes. GM130 (Golgi matrix protein of 130kDa) is a protein isolated from the Triton™ X-100-insoluble Golgi matrix and peripherally associated with the cis compartment, as demonstrated by co-localization with syntaxin5. GM130 is homologous to the Golgi autoantigen golgin 95. GM130 interacts through its N-terminal domain with p115 and with the Golgi membranes at the C-terminal portion. Furthermore, the mitotic phosphorylation of GM130 blocks the interaction with p115. Thus, GM130 appears to function as a structural element of the Golgi apparatus that also provides attachment sites for membranes and other Golgi proteins. The 35/GM130 monoclonal antibody recognizes GM130, regardless of phosphorylation status.
Host Species: Mouse
Clone: 35
Isotype: IgG1
Species Reactivity [for Features Main]: Human
Immunogen: Rat GM130 aa. 869-982
Immunofluorescence, Immunoprecipitation
Specifications
| GM130 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified |
| 35/GM130 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Canine, Human, Mouse, Rat | |
| Rat GM130 aa. 869-982 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.