missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSF Rat anti-Mouse, Unlabeled, Clone: MP1-22E9, BD
Rat Monoclonal Antibody
Brand: BD Biosciences 554404
This item is not returnable.
View return policy
Description
The MP1-22E9 antibody reacts with mouse granulocyte/macrophage-colony stimulating factor (GM-CSF). The immunogen used to generate the MP1-22E9 hybridoma was yeast-expressed recombinant mouse GM-CSF. This is a neutralizing antibody.
Specifications
| GM-CSF | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant mouse GM-CSF | |
| 0.5 mg | |
| Immunology | |
| Mouse | |
| IgG2a |
| ELISA | |
| MP1-22E9 | |
| Unconjugated | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction