Learn More
GYS2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542763
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: TLSRAFPDKF HVELTSPPTT EGFKYPRPSS VPPSPSGSQA SSPQSSDVED Sequence homology: Cow: 93%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%
GYS2 Polyclonal antibody specifically detects GYS2 in Human samples. It is validated for Western Blot.Specifications
GYS2 | |
Polyclonal | |
Unconjugated | |
GYS2 | |
BC021322; cb765; Glycogen [starch] synthase, liver; glycogen liver; glycogen synthase; glycogen synthase 2; glycogen synthase 2 (liver); glycogen synthase, liver; GLYSN; GYS2; LGS; zgc:112057 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2998 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P54840 | |
GYS2 | |
Synthetic peptide directed towards the middle region of human GYS2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |