Learn More
GK2 Monoclonal Antibody (3G4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518967
Description
Peptide Sequence: MAAPKTAAVG PLVGAVVQGT NSTRFLVFNS KTAELLSHHK VELTQEFPKE GWVEQDPKEI LQSVYECIAR TCEKLDELNI DISNIKAVGV SNQRETTVIW DKLTGEPLYN AVVWLDLRTQ TTVEDLSKKI PGNSNFVKSK TGLPLSTYFS AVKLRWMLDN VRNVQKAVEE GRALFGTIDS WLIWSLTGGV NGGVHCTDVT NASRTMLFNI HSLEWDKELC DFFEIPMDLL PNVFSSSEIY GLIKTGALEG VPISGCLGDQ CAALVGQMCF QEGQAKNTYG TGCFLLCNTG RKCVFSEHGL LTTVAYKLGR EKPAYYALEG SVAIAGAVIR WLRDNLGIIE TSGDIERLAK EVGTSYGCYF VPAFSGLYAP YWEPSARGIL CGLTQFTNKC HIAFAALEAV CFQTREILEA MNRDCGIPLR HLQVDGGMTN NKVLMQLQAD ILHIPVIKPF MPETTALGAA MAAGAAEGVS VWSLEPQALS VLRMERFEPQ IQATESEIRY ATWKKAVMKS MGWVTSQSPE GGDPSIFSSL PLGFFIVSSM VMLIGARYIS GVP
The human glycerol kinase gene family consists of at least 3 expressed loci. The GK1 locus on Xp21.3 is the site of mutations (deletions) causing glycerol kinase deficiency. It comprises 19 exons and is probably ancestral to several other genes which, because they are intronless, are suspected of having arisen by reverse transcriptase mediated events. These include 2 genes on chromosome 4. They are expressed as a single mRNA species in testis where expression is at a high level.
Specifications
| Glycerol kinase 2 | |
| Monoclonal | |
| 0.2-1.0 mg/mL | |
| PBS with no preservative; pH 7.4 | |
| Q14410 | |
| GK2 | |
| GK2 (AAH29820, 1 a.a. ∽ 553 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 3G4 | |
| Unconjugated | |
| GK2 | |
| GKP2, GKTA, ATP:glycerol 3-phosphotransferase 2, GK 2, glycerokinase 2, glycerol kinase pseudogene 2, glycerol kinase testis specific 2, glycerol kinase, testis specific 2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2712 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.