missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Glutamate Receptor 1 (AMPA subtype), Polyclonal, Abcam™
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Human, Marmoset (common). Predicted to work with Monkey
Immunogen: Synthetic peptide derived from within residues 850 to the C-terminus of Human Glutamate Receptor 1 (AMPA subtype).
Applications:
Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
Specifications
| Antigen | Glutamate Receptor 1 (AMPA subtype) |
| Classification | Polyclonal |
| Concentration | 0.3-0.9mg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from within residues 850 to the C-terminus of Human Glutamate Receptor 1 (AMPA subtype). |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Research Discipline | Neuroscience |
| Primary or Secondary | Primary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?