missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GLUT1 Polyclonal Antibody, Invitrogen™
Description
PA5-16793 targets GLUT-1 in IHC (P), WB, and IF applications and shows reactivity with Human and Rat samples. The PA5-16793 immunogen is a synthetic peptide derived from C-terminal of human GLUT-1.
Glucose is fundamental to the metabolism of mammalian cells. Several glucose transporter protein (Glut) isoforms have been identified and shown to function in response to insulin and IGF-1 induced signaling. GLUT-1 is detectable in many human tissues including those of the colon, lung, stomach, esophagus, and breast. GLUT-1 immunoreactivity in some cancers, including trans carcinoma of the urinary bladder, has been associated with aggressive behavior.
Specifications
Specifications
| Antigen | GLUT1 |
| Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.1% sodium azide; pH 7.4 |
| Gene | SLC2A1 |
| Gene Accession No. | P11166, P11167 |
| Gene Alias | GLUT-1, glucose transporter type 1, erythrocyte/brain, hepG2 glucose transporter, solute carrier family 2, facilitated glucose transporter member 1 |
| Gene Symbols | SLC2A1 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?