missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GLAST Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534197
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Mouse, Hamster, Elephant, Panda, Horse (93%) Marmoset, Rat, Pig (87%).
This gene encodes a member of a member of a high affinity glutamate transporter family. Mutations in this gene are associated with episodic ataxia, Type 6. Alternative splicing results in multiple transcript variants.
Specifications
| GLAST | |
| Polyclonal | |
| Unconjugated | |
| SLC1A3 | |
| AI504299; B430115D02Rik; EA6; Eaat1; EAAT-1; Excitatory amino acid transporter 1; FLJ25094; GLAST; GLAST1; GLAST-1; Glial glutamate transporter; glial high affinity glutamate transporter; GLU-T; GluT-1; glutamate transporter; glutamate/aspartate transporter; Gmt1; High-affinity neuronal glutamate transporter; MGluT1; Slc1a3; Sodium-dependent glutamate/aspartate transporter 1; solute carrier family 1 (glial high affinity glutamate transporter), member 3; solute carrier family 1 member 3; solute carrier family 1, member 3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6507 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 0.1% sodium azide | |
| P43003 | |
| SLC1A3 | |
| Synthetic 15 amino acid peptide from cytoplasmic domain of human SLC1A3 / GLAST. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction