Learn More
GluR6/GluR7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532427
Code : PN
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine, chicken, canine, amphibian, mouse, rabbit and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is brain tissue.
Glutamate receptors constitute the principal excitatory neurotransmitter receptors in brain. Two classes of glutamate receptors exist: ionotropic receptors, and metabotropic receptors. Ionotropic glutamate receptors are oligomeric complexes of various subunits (GluR1-7, NMDA1-3, KAI-2), which comprise ligand-gated calcium channels. Metabotropic glutamate receptors are G-protein coupled receptors, which when activated can activate phopholipases or adenylylcyclase, depending on the neuron.Specifications
GluR6/GluR7 | |
Polyclonal | |
GRIK3 | |
GRIK2, GRIK3, EAA4, GLR6, MRT6, GLUK6, GLUR6, GluK2, Glutamate Receptor 6/7, Glutamate Receptor 6, Glutamate Receptor 7 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2898, 2899, 54257 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
P42260, Q13002, Q13003 | |
GRIK2, GRIK3 | |
Synthetic peptide from C-terminus of human GluR6/7. | |
500 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |