Learn More
GluR δ2 Mouse, Unlabeled, Clone: 48, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 611006
Descripción
Glutamate is a major excitatory neurotransmitter of the CNS. The diversity of glutamate is exemplified by two distinct groups of receptors: ionotropic and metabotropic. Ionotroic receptors are ligand-gated cation channels. They can be subdivided into two classes: NMDA and AMPA/kainate receptors. GluR δ2 exhibits only 25% amino acid identity to either ionotropic receptor type. It presents selective and abundant exression in cerebellar Purkinje cells. It is targeted to a subset of Purkinje cell spines and is coexpressed with ionotropic receptors. GluR δ2 is involved in motor coordination, Purkinje cell synapse formation, and cerebellar long-term depression (LTD). The latter is a decrease in the efficacy of the synaptic transmission between parallel fibers and Purkinje neurons and is a cellular basis of motor learning. In fact, mGluR δ2-deficient mice lack LTD. Additionally, an amino acid substitution in transmembrane III of δ2 is responsible for the neurodegeneration seen in Lurcher mice. This substitution is a gain of function mutation that results in disruption of Purkinje membrane potential. Thus, GluR δ2 is an important regulatory component of the Purkinje GluR channel.
Immunofluorescence, Western Blotting
Especificaciones
| GluR δ2 | |
| Monoclonal | |
| 250μg/mL | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Mouse GluR δ2 aa. 665-786 | |
| 50 μg | |
| Cell Biology | |
| Mouse, Rabbit, Rat | |
| IgG1 |
| Western Blot | |
| 48 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.