missing translation for 'onlineSavingsMsg'
Learn More

anti-Glucose Transporter GLUT3, Polyclonal, Abcam™

Brand:  Abcam AB41525-100UG

Encompass

Product Code. 13058715

  • £615.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Rat, Human
Immunogen: Synthetic peptide from the C terminus of Glucose Transporter GLUT3.

Applications:

Dot Blotting, ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Specifications

Specifications

Glucose Transporter GLUT3
0.5mg/mL
Rabbit
Affinity Purified
Cancer and Tumor Biology, Cell Biology, Signal Transduction
Human, Mouse, Rat
Polyclonal
Unconjugated
Synthetic peptide from the C terminus of Glucose Transporter GLUT3.
100 μg
Primary
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.