Learn More
anti-Glucagon Receptor, Polyclonal, Abcam™
Brand: Abcam AB75240-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Human. Predicted to work with Mouse, Rat
Immunogen: Synthesized peptide derived from internal of human Glucagon Receptor
Applications:
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
Specifications
| Glucagon Receptor | |
| 1mg/mL | |
| Rabbit | |
| Affinity Purified | |
| Cancer and Tumor Biology, Signal Transduction | |
| Human, Mouse, Rat |
| Polyclonal | |
| Unconjugated | |
| Synthesized peptide derived from internal of human Glucagon Receptor | |
| 100 μg | |
| Primary | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.