Learn More
GLO1, Mouse, Clone: 4C12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GLO1.
Brand: Abnova H00002739-M01.100ug
Description
The enzyme encoded by this gene is responsible for the catalysis and formation of S-lactoyl-glutathione from methylglyoxal condensation and reduced glutathione. Glyoxalase I is linked to HLA and is localized to 6p21.3-p21.1, between HLA and the centromere. [provided by RefSeq
Sequence: MAEPQPPSGGLTDEAALSYCSDADPSTKDFLLQQTMLRVKDPKKSLDFYTRVLGMTLIQKCDFPIMKFSLYFLAYEDKNDIPKEKDEKIAWALSRKATSpecifications
| GLO1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC011365 | |
| GLO1 | |
| GLO1 (AAH11365.1, 1 a.a. ∼ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA | |
| 4C12 | |
| Mouse monoclonal antibody raised against a partial recombinant GLO1. | |
| GLO1 | |
| GLOD1/GLYI | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2739 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.