Learn More
GLE1, Mouse, Clone: 1D8, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GLE1.
Brand: Abnova H00002733-M03.100ug
Description
This gene encodes a predicted 75kDa polypeptide with high sequence and structure homology to yeast Gle1p, which is nuclear protein with a leucine-rich nuclear export sequence essential for poly(A)+RNA export. Inhibition of human GLE1L by microinjection of antibodies against GLE1L in HeLa cells resulted in inhibition of poly(A)+RNA export. Immunoflourescence studies show that GLE1L is localized at the nuclear pore complexes. This localization suggests that GLE1L may act at a terminal step in the export of mature RNA messages to the cytoplasm. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: RMKGTEGLRLWQEEQERKVQALSEMASEQLKRFDEWKELKQHKEFQDLREVMEKSSREALGHQEKLKAEHRHRAKILNLKLREAEQQRVKQAEQERLRKEESpecifications
| GLE1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001003722 | |
| GLE1 | |
| GLE1 (NP_001003722.1, 140 a.a. ∼ 240 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunofluorescence, Western Blot | |
| 1D8 | |
| Mouse monoclonal antibody raised against a partial recombinant GLE1. | |
| GLE1 | |
| GLE1L/LCCS/LCCS1/hGLE1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2733 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.