Learn More
GINS2 Monoclonal Antibody (3H4)
GREENER_CHOICE
GINS2 Monoclonal Antibody (3H4)
Brand:  Invitrogen MA521970
Product Code. 15539062
  • £442.00 / 100µg
Specifications
GINS2
Monoclonal
0.5 mg/mL
PBS with no preservative; pH 7.4
Q9Y248
GINS2
GINS2 (AAH03186, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA, Western Blot
3H4
Unconjugated
GINS2
2210013I18Rik; 4833427B12Rik; AI323585; CGI-122; DC5; DNA replication complex GINS protein PSF2; GINS complex subunit 2; GINS complex subunit 2 (Psf2 homolog); Gins2; HSPC037; Pfs2; PSF2; RGD1311055
Mouse
Affinity Chromatography
RUO
51659
-20°C, Avoid Freeze/Thaw Cycles