Learn More
golgi associated, gamma adaptin ear containing, ARF binding protein 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human GGA1 protein.
Brand: Abnova H00026088-D01P.100ug
Description
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) protein family. Members of this family are ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MEPAMEPETLEARINRATNPLNKELDWASINGFCEQLNEDFEGPPLATRLLAHKIQSPQEWEAIQALTVLETCMKSCGKRFHDEVGKFRFLNELIKVVSPKYLGSRTSEKVKNKILELLYSWTVGLPEEVKIAEAYQMLKKQGIVKSDPKLPDDTTFPLPPPRPKNVIFEDEEKSKMLARLLKSSHPEDLRAANKLIKEMVQEDQKRMEKISKRVNAIEEVNNNVKLLTEMVMSHSQGGAAAGSSEDLMKELYQRCERMRPTLFRLASDTEDNDEALGLSDPTPPSGPSLDGTGWNSFQSSDATEPPAPALAQAPSMESRPPAQTSLPASSGLDDLDLLGKTLLQQSLPPESQQVRWEKQQPTPRLTLRDLQNKSSSCSSPSSSATSLLHTVSPEPPRPPQQPVPTELSLASITVPLESIKPSNILPVTVYDQHGFRILFHFARDPLPGRSDVLVVVVSMLSTAPQPIRNIVFQSAVPKVMKVKLQPPSGTELPAFNPIVHPSAITQVLLLANPQKEKVRLRYKLTFTMGDQTYNEMGDVDQFPPPETWGSLSpecifications
| golgi associated, gamma adaptin ear containing, ARF binding protein 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human GGA1 protein. | |
| GGA1 | |
| GGA1 | |
| GGA1 (NP_001001560.1, 1 a.a. ∼ 552 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001001560 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 26088 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.