Learn More
golgi associated, gamma adaptin ear containing, ARF binding protein 1, Mouse, Clone: 1G8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant GGA1.
Brand: Abnova H00026088-M02.100ug
Description
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) protein family. Members of this family are ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MEPAMEPETLEARINRATNPLNKELDWASINGFCEQLNEDFEGPPLATRLLAHKIQSPQEWEAIQALTVRRGEATIRPPPCDDTKGGQDSpecifications
| golgi associated, gamma adaptin ear containing, ARF binding protein 1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC029388 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| Antibody |
| ELISA, KnockDown, Western Blot | |
| 1G8 | |
| Mouse monoclonal antibody raised against a full length recombinant GGA1. | |
| GGA1 | |
| GGA1 | |
| GGA1 (AAH29388, 1 a.a. ∼ 89 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Yes | |
| 26088 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.