missing translation for 'onlineSavingsMsg'
Learn More

anti-GFAP, Clone: 2A5, Abcam™

Product Code. 13031076
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
13031076 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13031076 Supplier Abcam Supplier No. AB4648100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.

Host Species: Mouse
Clone: 2A5
Isotype: IgG1, κ
Cross Reactivity: Mouse, Rat, Cow, Human, Pig. Predicted to work withall Mammals. Does not react with Chicken.
Immunogen: A preparation of purified pig spinal core GFAP.

Applications:

ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen GFAP
Classification Monoclonal
Clone 2A5
Conjugate Unconjugated
Host Species Mouse
Immunogen A preparation of purified pig spinal core GFAP.
Quantity 100 μL
Research Discipline Neuroscience, Signal Transduction, Stem Cell Biology, Tags And Cell Markers
Primary or Secondary Primary
Target Species Bovine, Human, Mammalia, Mouse, Pig, Rat
Form Supernatant
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.