Learn More
Invitrogen™ Gentamicin Monoclonal Antibody (GB26.16)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA110726
Product Code. 11518992
  • £479.00 / 500µg
Description
MA1-10726 detects Gentamicin. MA1-10726 has been successfully used in ELISA applications. The MA1-10726 immunogen is Gentamicin, conjugated to KLH.
Gentamicin is an aminoglycoside antibiotic, used to treat many types of bacterial infections, particularly those caused by Gram-negative bacteria. However, gentamicin is not used for Neisseria gonorrhoeae, Neisseria meningitidis or Legionella pneumophila bacterial infections (because of the risk of the patient going into shock from lipid A endotoxin found in certain gram negative organisms).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Gentamicin
Monoclonal
1.1 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein G
RUO
Chemical
Antibody
IgG2a
ELISA
GB26.16
Unconjugated
Gentacycol; Garamycin; Gentavet; Gentamicinum; C21H43N5O7
Gentamicin, conjugated to KLH.
500 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Chemical