Learn More
anti-GEMIN6 Matched Antibody Pair, Abnova
Brand: Abnova H00079833-AP21.1Set
Includes: Rabbit MaxPab™ affinity purified polyclonal anti-GEMIN6 (100μg); Mouse purified polyclonal anti-GEMIN6 (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
GEMIN6 is part of a large macromolecular complex, localized to both the cytoplasm and the nucleus, that plays a role in the cytoplasmic assembly of small nuclear ribonucleoproteins (snRNPs). Other members of this complex include SMN (MIM 600354), GEMIN2 (SIP1; MIM 602595), GEMIN3 (DDX20; MIM 606168), GEMIN4 (MIM 606969), and GEMIN5 (MIM 607005).
ELISA
Specifications
| GEMIN6 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GEMIN6. | |
| FLJ23459 | |
| Mouse, Rabbit | |
| 1 Set | |
| Stem Cell Biology | |
| 79833 | |
| Human |
| ELISA | |
| Unconjugated | |
| GEMIN6 | |
| GEMIN6 | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-GEMIN6 (100 μg) 2. Detection antibody: mouse purified polyclonal anti-GEMIN6 (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.