missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gemin 4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519576
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and cow based on sequence homology.
The product of this gene is part of a large complex localized to the cytoplasm, nucleoli, and to discrete nuclear bodies called Gemini bodies . The complex functions in spliceosomal snRNP assembly in the cytoplasm, and regenerates spliceosomes required for pre-mRNA splicing in the nucleus. The encoded protein directly interacts with a DEAD box protein and several spliceosome core proteins. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
| Gemin 4 | |
| Polyclonal | |
| Unconjugated | |
| P57678 | |
| GEMIN4 | |
| Synthetic peptide conjugated to KLH derived from within residues 250 - 350 of Human Gemin 4. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| GEMIN4 | |
| DKFZp434B131, DKFZp434D174, HC56, HCAP1, HHRF-1, p97, HCC-associated protein 1; component of gems 4; gemin-4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 50628 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction