Learn More
GDF2, Mouse, Clone: 4D2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GDF2.
Brand: Abnova H00002658-M01.100ug
Description
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in rodents suggest that this protein plays a role in the adult liver and in differentiation of cholinergic central nervous system neurons. [provided by RefSeq
Sequence: SAGAGSHCQKTSLRVNFEDIGWDSWIIAPKEYEAYECKGGCFFPLADDVTPTKHAIVQTLVHLKFPTKVGKACCVPTKLSPISVLYKDDMGVPTLKYHYESpecifications
| GDF2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_016204 | |
| GDF2 | |
| GDF2 (NP_057288, 320 a.a. ∼ 419 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA | |
| 4D2 | |
| Mouse monoclonal antibody raised against a partial recombinant GDF2. | |
| GDF2 | |
| BMP-9/BMP9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2658 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.