Learn More
K(lysine) acetyltransferase 2A, Mouse, Clone: 4D3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GCN5L2.
Brand: Abnova H00002648-M01.100ug
Description
KAT2A, or GCN5, is a histone acetyltransferase (HAT) that functions primarily as a transcriptional activator. It also functions as a repressor of NF-kappa-B (see MIM 164011) by promoting ubiquitination of the NF-kappa-B subunit RELA (MIM 164014) in a HAT-independent manner (Mao et al., 2009 [PubMed 19339690]).[supplied by OMIM
Sequence: KNLLAQIKSHPSAWPFMEPVKKSEAPDYYEVIRFPIDLKTMTERLRSRYYVTRKLFVADLQRVIANCREYNPPDSEYCRCASALEKFFYFKLKEGGLIDKSpecifications
| K(lysine) acetyltransferase 2A | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC032743 | |
| KAT2A | |
| GCN5L2 (AAH32743, 738 a.a. ∼ 837 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 4D3 | |
| Mouse monoclonal antibody raised against a partial recombinant GCN5L2. | |
| KAT2A | |
| GCN5/GCN5L2/MGC102791/PCAF-b/hGCN5 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 2648 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.