missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNB3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSG00021W
This item is not returnable.
View return policy
Description
Glycerol (1:1) may be added for additional stability.
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. A single-nucleotide polymorphism (C825T) in this gene is associated with essential hypertension and obesity. This polymorphism is also associated with the occurrence of the splice variant GNB3-s, which appears to have increased activity. GNB3-s is an example of alternative splicing caused by a nucleotide change outside of the splice donor and acceptor sites. Additional splice variants may exist for this gene, but they have not been fully described.
Specifications
| GNB3 | |
| Polyclonal | |
| GNB3 | |
| Transducin beta chain 3, GNB3, Guanine nucleotide binding protein subunit beta-3 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| P16520, Q61011, P52287 | |
| GNB3 | |
| A synthetic peptide from aa region 2-50 of human GNB3 conjugated to an immunogenic carrier protein was used as the antigen | |
| RUO | |
| 14695, 2784, 60449 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction