Learn More
Gaussia luciferase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1181
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Western blot analysis of PA1-181 detects a ∽20 kDa protein in the supernatant of Gaussia luciferase-secreting cells. PA1-181 can be used for activity-independent verification of Gaussia luciferase expression.
Gaussia luciferase Polyclonal antibody specifically detects Gaussia luciferase in Luciferase samples. It is validated for Western Blot.Specifications
Gaussia luciferase | |
Polyclonal | |
Unconjugated | |
Gaussia luciferase | |
Purified recombinant Gaussia luciferase expressed in E. coli | |
100 μg | |
Primary | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
Rabbit | |
Protein G | |
RUO | |
Luciferase | |
Antibody | |
IgG |