missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GATA3 Mouse, Alexa Fluor 647, Clone: L50-823, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560068
This item is not returnable.
View return policy
Description
GATA3 (GATA binding protein 3) is a member of the GATA family of transcription factors. This ∼50-kDa nuclear protein regulates the development and subsequent maintenance of multiple tissues. GATA3 is involved in the development of T lymphocytes (regulates T cell receptor subunit gene expression) and the differentiation of mature T cells to become Th2 cells. The expressed levels of normal or mutant GATA3 are also associated with the behaviors of various cancer cells including estrogen receptor-positive breast carcinoma cells.
Specifications
| GATA3 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human, Rat | |
| IgG1 κ |
| Flow Cytometry | |
| L50-823 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Conserved peptide between the trans-activation and DNA-binding domains of human, mouse and rat GATA3 | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction