Learn More
GAPDH Loading Control Monoclonal Antibody (GA1R), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA5157381MG
Code : NEW
Additional Details : Weight : 0.01000kg
Description
This antibody detects GAPDH from BL-21 bacteria, Sf9 insect, Saccharomyces cerevisiae (yeast), human, mouse, rat, rabbit, hamster, and chicken samples. MA5-15738 has been successfully used in Western blot, ICC, IF, IHC (P), FACS and ELISA. MA5-15738 may be stored at 4oC short term. Add 0.05% sodium azide if desired. For long term storage, store at -20oC, avoiding freeze/thaw cycles.
GAPDH Loading Control Monoclonal antibody specifically detects GAPDH Loading Control in Bacteria, Canine, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
GAPDH Loading Control | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
O14556, P00356, P04406, P04797, P16858, P17244, P46406, Q28259, Q64467, Q9ESV6 | |
GAPDH, GAPDHS | |
Recombinant GAPDH | |
1 mg | |
Primary | |
Bacteria, Canine, Chicken, Hamster, Human, Insect, Mouse, Rabbit, Rat, Yeast | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
GA1R | |
Unconjugated | |
GAPDHS | |
38 kDa BFA-dependent ADP-ribosylation substrate; aging-associated gene 9 protein; BARS-38; bb02e05; cb350; cb609; CDABP0047; EC 1.2.1.12; epididymis secretory protein Li 278; epididymis secretory sperm binding protein Li 162eP; fb71f08; fk58c09; G3PD; G3PDH; GAPD; GAPD2; gapdh; GAPDH2; GAPDH-2; GAPDHS; Gapds; Gapd-s; glceraldehyde-3-phosphate dehydrogenase; glyceraldehyde 3-phosphate dehydrogenase; glyceraldehyde 3-phosphate dehydrogenase, testis-specific; glyceraldehyde phosphate dehydrogenase; glyceraldehyde-3-phosphate dehydrogenase; glyceraldehyde-3-phosphate dehydrogenase (G3PDH); glyceraldehyde-3-phosphate dehydrogenase 2; glyceraldehyde-3-phosphate dehydrogenase GAPDH; glyceraldehyde-3-phosphate dehydrogenase like-17 protein; glyceraldehyde-3-phosphate dehydrogenase type 2; glyceraldehyde-3-phosphate dehydrogenase, spermatogenic; glyceraldehyde-3-phosphate dehydrogenase, testis-specific; glyceraldehyde-phosphate-dehydrogenase; glycerine aldehyde 3-phosphate dehydrogenase; HEL-S- | |
Mouse | |
Protein A | |
RUO | |
100009074, 100352213, 100736557, 100767862, 14433, 14447, 24383, 2597, 26330, 374193, 403755, 476483, 66020 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |