missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma Adaptin Monoclonal Antibody (100/3), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA125066
This item is not returnable.
View return policy
Description
Recommended positive controls: bovine brain. MA1-25066 will not react with mouse or rat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
gamma Adaptin Monoclonal antibody specifically detects gamma Adaptin in Bovine, Human, Non-human Primate samples. It is validated for Immunocytochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot.Specifications
gamma Adaptin | |
Monoclonal | |
0.9 mg/mL | |
Ascites with 0.1% sodium azide | |
AA409002; adapter-related protein complex 1 subunit gamma-1; adaptor protein complex AP-1 gamma 1 subunit; adaptor protein complex AP-1 subunit gamma-1; adaptor protein complex AP-1, gamma 1 subunit; adaptor related protein complex 1 gamma 1 subunit; adaptor related protein complex 1 subunit gamma 1; adaptor-related protein complex 1 gamma 1 subunit; adaptor-related protein complex 1 subunit gamma-1; adaptor-related protein complex 1, gamma 1 subunit; ADTG; AP-1 complex subunit gamma-1; Ap1g1; AU041323; AW551707; CLAPG1; clathrin assembly protein complex 1 gamma large chain; clathrin assembly protein complex 1 gamma-1 large chain; clathrin-associated/assembly/adaptor protein, large, gamma 1; D8Ertd374e; gamma adaptin; Gamma1-adaptin; gamma-adaptin; golgi adaptor HA1/AP1 adaptin gamma subunit; golgi adaptor HA1/AP1 adaptin subunit gamma-1; MGC18255; testicular tissue protein Li 21 | |
Mouse | |
100 μL | |
Primary | |
Bovine, Human, Non-human Primate | |
Antibody | |
IgG2b |
Immunocytochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot | |
100/3 | |
Unconjugated | |
AP1G1 | |
AP1G1 | |
AP-1 adaptor from bovine brain | |
RUO | |
507677 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |