missing translation for 'onlineSavingsMsg'
Learn More
Learn More
galactokinase 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant GALK1.
Brand: Abnova H00002584-A02.50uL
This item is not returnable.
View return policy
Description
Galactokinase is a major enzyme for the metabolism of galactose and its deficiency causes congenital cataracts during infancy and presenile cataracts in the adult population. [provided by RefSeq
Sequence: PCGIMDQFISLMGQKGHALLIDCRSLETSLVPLSDPKLAVLITNSNVRHSLASSEYPVRRRQCEEVARALGKESLREVQLEELEAARDLVSKEGFRRARSpecifications
| galactokinase 1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant GALK1. | |
| GALK1 | |
| GALK/GK1 | |
| Mouse | |
| 50 μL | |
| Yes | |
| 2584 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_000154 | |
| GALK1 | |
| GALK1 (NP_000145, 181 a.a. ∼ 279 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction