missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GABPB2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA546860
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: PDGQQVLTVP ATDIAEETVI SEEPPAKRQC IEIIENRVES AEIEEREALQ Sequence homology: Chicken: 92%
GABPB2 Polyclonal antibody specifically detects GABPB2 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
GABPB2 | |
Polyclonal | |
Unconjugated | |
Gabpb2 | |
1810015F01Rik; 5830427M07; 9430006E19Rik; A430024B14Rik; AV050852; BABPB2; E4TF1; E4TF1-47; E4TF1-53; E4TF1B; GA binding protein transcription factor beta subunit 1; GA binding protein transcription factor beta subunit 1 transcript variant gamma-2; GA binding protein transcription factor beta subunit 2; GA binding protein transcription factor subunit beta 2; GA binding protein transcription factor, alpha subunit pseudogene; GA binding protein transcription factor, beta subunit 1; GA binding protein transcription factor, beta subunit 2; GA repeat binding protein, beta 2; GA-binding protein beta-2-1; GA-binding protein subunit beta-1; GA-binding protein subunit beta-2; GABP subunit beta-1; GABP subunit beta-2; GABP subunit beta-2-1; GABP2; GABPAP; GABPB; GABPB1; GABPB-1; Gabpb2; GABPB-2; Gabpb2-1; LOW QUALITY PROTEIN: GA-binding protein subunit beta-2; NRF2B1; NRF2B2; Nuclear respiratory factor 2; RGD1565234; RP11-68I18.1; Transcription factor E4TF1-47; transcription factor E4TF1-53 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
126626 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8TAK5 | |
Gabpb2 | |
Synthetic peptide directed towards the C-terminal of human GABPB2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |