Learn More
GABBR1, Mouse, Clone: 2D7, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GABBR1.
Brand: Abnova H00002550-M01.100ug
Description
Gamma-aminobutyric acid (GABA) is the main inhibitory neurotransmitter in the mammalian central nervous system. GABA exerts its effects through ionotropic [GABA(A/C)] receptors, to produce fast synaptic inhibition, and metabotropic [GABA(B)] receptors, to produce slow, prolonged inhibitory signals. The GABA(B) receptor consists of a heterodimer of two related 7-transmembrane receptors, GABA(B) receptor 1 and GABA(B) receptor 2. The GABA(B) receptor 1 gene is mapped to chromosome 6p21.3 within the HLA class I region close to the HLA-F gene. Susceptibility loci for multiple sclerosis, epilepsy, and schizophrenia have also been mapped in this region. Alternative splicing of this gene generates multiple transcript variants. [provided by RefSeq
Sequence: AVYIGALFPMSGGWPGGQACQPAVEMALEDVNSRRDILPDYELKLIHHDSKCDPGQATKYLYELLYNDPIKIILMPGCSSVSTLVAEAARMWNLIVLSYGSpecifications
| GABBR1 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC050532 | |
| GABBR1 | |
| GABBR1 (AAH50532, 52 a.a. ∼ 151 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunofluorescence, KnockDown, Western Blot | |
| 2D7 | |
| Mouse monoclonal antibody raised against a partial recombinant GABBR1. | |
| GABBR1 | |
| FLJ92613/GABAB(1e)/GABABR1/GABBR1-3/GPRC3A/dJ271M21.1.1/dJ271M21.1.2/hGB1a | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2550 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.