missing translation for 'onlineSavingsMsg'
Learn More

Gαq Mouse, Unlabeled, Clone: 10, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612705

 View more versions of this product

Product Code. 15826269

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

The GTP binding regulatory proteins (G proteins) consist of three subunits: α, β, and γ. These heterotrimeric proteins function at membranes to relay signals from cell surface receptors to intracellular effectors. The α subunit is unique for each G protein and contains the site of GTP binding and hydrolysis, as well sites for receptor and effector interactions. The βγ subunit complex interacts directly with receptors and the α subunit. The Gα family includes four families: the Gαs family including Gαs , Gαo1f, and Gαt, the Gαi family including Gαi, Gαo, and Gαz, the Gαq/Gα11 family and the Gα12/13 family. The Gαq protein is 88% homologus with Gα11 and both are widely expressed. These G proteins activate phospholipase C proteins, which induce calcium signaling events. G protein coupled receptors (GPCRs) involved in regulating Wnt signaling activate Gαq, phospholipase Cβ, and induce calcium-dependent activation of calpain. These events promote β-catenin nuclear export and proteolysis. Gαq has also been implicated in metabotropic glutamate receptor signaling. Thus, Gαq isoforms activate phospholipase C proteins in various G-protein coupled receptor pathways.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Gαq
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human Gαq aa. 22-31
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
10
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.