missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPRC6A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132735
This item is not returnable.
View return policy
Description
PA1-32735 detects GPCR GPRC6A from human samples.
GPRC6A is an Orphan-U GPCR with an unknown ligand. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This "e;turn-off"e; activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.
Specifications
| GPRC6A | |
| Polyclonal | |
| Unconjugated | |
| GPRC6A | |
| bA86F4.3; G protein-coupled receptor class C group 6 member A; G protein-coupled receptor, class C, group 6, member A; G protein-coupled receptor, family C, group 6, member A; GPCR; Gprc6a; G-protein coupled receptor family C group 6 member A; G-protein coupled receptor GPCR33; hGPCR33; hGPRC6A; predicted with SOSUI analysis; seven transmembrane helix receptor | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 222545 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q5T6X5 | |
| GPRC6A | |
| Synthetic 17 amino acid peptide from N-terminal extracellular domain of human GPRC6A | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido