missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR133 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA132849
This item is not returnable.
View return policy
Description
PA1-32849 detects GPCR GPR133 from human samples.
GPRs are characterized by the presence of 7 transmembrane domains. GPR133 is an orphan receptor.
Specifications
| GPR133 | |
| Polyclonal | |
| Unconjugated | |
| ADGRD1 | |
| Adgrd1; adhesion G protein-coupled receptor D1; adhesion G-protein coupled receptor D1; DKFZp434B1272; E230012M21Rik; FLJ16770; G protein-coupled receptor 133; GPR133; G-protein coupled receptor 133; G-protein coupled receptor PGR25; MGC138512; MGC138514; Pgr25; probable G-protein coupled receptor 133 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283383 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q6QNK2 | |
| ADGRD1 | |
| Synthetic 20 amino acid peptide from N-terminal extracellular domain of human GPR133 | |
| 25 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction