Learn More
FXN, Mouse, Clone: 1D9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FXN.
Brand: Abnova H00002395-M01.100ug
Description
This nuclear gene encodes a mitochondrial protein which belongs to FRATAXIN family. The protein functions in regulating mitochondrial iron transport and respiration. The expansion of intronic trinucleotide repeat GAA results in Friedreich ataxia. Alternative splicing results in multiple transcript variants. [provided by RefSeq
Sequence: DETTYERLAEETLDSLAEFFEDLADKPYTFEDYDVSFGSGVLTVKLGGDLGTYVINKQTPNKQIWLSSPSSGPKRYDWTGKNWVYSHDGVSLHELLAAELTKALKTKLDLSpecifications
| FXN | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_000144 | |
| FXN | |
| FXN (AAH48097.1, 91 a.a. ∼ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
| ELISA, Immunoprecipitation, Western Blot | |
| 1D9 | |
| Mouse monoclonal antibody raised against a partial recombinant FXN. | |
| FXN | |
| CyaY/FA/FARR/FRDA/MGC57199/X25 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2395 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.