Learn More
ferritin, heavy polypeptide 1, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human FTH1 protein.
Brand: Abnova H00002495-D01P.100ug
Description
This gene encodes the heavy subunit of ferritin, the major intracellular iron storage protein in prokaryotes and eukaryotes. It is composed of 24 subunits of the heavy and light ferritin chains. Variation in ferritin subunit composition may affect the rates of iron uptake and release in different tissues. A major function of ferritin is the storage of iron in a soluble and nontoxic state. Defects in ferritin proteins are associated with several neurodegenerative diseases. This gene has multiple pseudogenes. Several alternatively spliced transcript variants have been observed, but their biological validity has not been determined. [provided by RefSeq
Sequence: MTTASTSQVRQNYHQDSEAAINRQINLELYASYVYLSMSYYFDRDDVALKNFAKYFLHQSHEEREHAEKLMKLQNQRGGRIFLQDIKKPDCDDWESGLNAMECALHLEKNVNQSLLELHKLATDKNDPHLCDFIETHYLNEQVKAIKELGDHVTNLRKMGAPESGLAEYLFDKHTLGDSDNESSpecifications
| ferritin, heavy polypeptide 1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human FTH1 protein. | |
| FTH1 | |
| FHC/FTH/FTHL6/MGC104426/PIG15/PLIF | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antibody |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002032 | |
| FTH1 | |
| FTH1 (NP_002023.2, 1 a.a. ∼ 183 a.a) full-length human protein. | |
| 100 μg | |
| Endocrine/Metabolism | |
| 2495 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.