Learn More
FPGT Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531546
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431. Predicted reactivity: Dog (87%), Pig (86%), Rabbit (84%), Rhesus Monkey (96%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
L-fucose is a key sugar in glycoproteins and other complex carbohydrates since it may be involved in many of the functional roles of these macromolecules, such as in cell-cell recognition. The fucosyl donor for these fucosylated oligosaccharides is GDP-beta-L-fucose. There are two alternate pathways for the biosynthesis of GDP-fucose; the major pathway converts GDP-alpha-D-mannose to GDP-beta-L-fucose. The protein encoded by this gene participates in an alternate pathway that is present in certain mammalian tissues, such as liver and kidney, and appears to function as a salvage pathway to reutilize L-fucose arising from the turnover of glycoproteins and glycolipids. This pathway involves the phosphorylation of L-fucose to form beta-L-fucose-1-phosphate, and then condensation of the beta-L-fucose-1-phosphate with GTP by fucose-1-phosphate guanylyltransferase to form GDP-beta-L-fucose.Specifications
FPGT | |
Polyclonal | |
Unconjugated | |
FPGT | |
1700016E03Rik; Fpgt; fucose-1-phosphate guanyltransferase; fucose-1-phosphate guanylyltransferase; GDP-beta-L-fucose pyrophosphorylase; GDP-L-fucose diphosphorylase; GDP-L-fucose pyrophosphorylase; GFPP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8790 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.94 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O14772 | |
FPGT | |
Recombinant fragment corresponding to a region within amino acids 33 and 326 of Human FPGT | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |