missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FOXR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530909
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Forkhead-box (FOX) transcription factors are implicated in carcinogenesis through gene amplification, retroviral integration, or chromosomal translocation. Human FOXN6 mRNA was expressed in breast cancer cell line and primary breast cancer.Specifications
FOXR2 | |
Polyclonal | |
Unconjugated | |
FOXR2 | |
EG436240; Forkhead box protein N6; forkhead box protein R2; forkhead box R2; Foxn6; Foxr2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
139628 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6PJQ5 | |
FOXR2 | |
Recombinant fragment corresponding to a region within amino acids 122 and 311 of Human FOXR2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |