missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FOXQ1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA540772
This item is not returnable.
View return policy
Description
Peptide sequence: MKLEVFVPRA AHGDKQGSDL EGAGGSDAPS PLSAAGDDSL GSDGDCAANS Sequence homology: Human: 100%; Mouse: 85%; Pig: 86%; Rat: 85%.
FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis.
Specifications
| FOXQ1 | |
| Polyclonal | |
| Unconjugated | |
| FOXQ1 | |
| forkhead box protein Q1; forkhead box Q1; FOXQ1; Hepatocyte nuclear factor 3 forkhead homolog 1; Hfh1; HFH-1; Hfh1l; HFH-1l; HNF-3/forkhead homolog 1 like; HNF-3/forkhead homolog-1; HNF-3/forkhead-like protein 1; sa; satin; winged helix/forkhead transcription factor | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 94234 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9C009 | |
| FOXQ1 | |
| Synthetic peptide directed towards the N-terminal of human FOXQ1 (aa 1-50). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction