Learn More
FOXO3A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527145
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, Mouse brain, C2C12, Jurkat, Raji. Predicted reactivity: Mouse (100%), Rat (100%), Pig (100%), Sheep (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.Specifications
FOXO3A | |
Polyclonal | |
Unconjugated | |
FOXO3 | |
1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; C76856; Fkhr2; FKHRL1; FKHRL1P2; forkhead box O3; forkhead box O3a; forkhead box O3A transcription factor; forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; forkhead in rhabdomyosarcoma-like 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO2; FOXO3; FOXO-3; FOXO3A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2309, 56484 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.56 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O43524, Q9WVH4 | |
FOXO3 | |
Synthetic peptide corresponding to a region within amino acids 611 and 673 of Human FOXO3A | |
100 μL | |
Primary | |
Human, Mouse, Rodent | |
Antibody | |
IgG |