Learn More
FOXF2, Mouse, Clone: 2F5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FOXF2.
Brand: Abnova H00002295-M06.100ug
Description
FOXF2 encodes forkhead box F2, one of many human homologues of the Drosophila melanogaster transcription factor forkhead. FOXF2 is expressed in lung and placenta, and has been shown to transcriptionally activate several lung-specific genes. [provided by RefSeq
Sequence: SPYLKQPPALTPSSNPAASAGLHSSMSSYSLEQSYLHQNAREDLSVGLPRYQHHSTPVCDRKDFVLNFNGISSFHPSASGSYYHHHHQSVCQDIKPCVSpecifications
| FOXF2 | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001452 | |
| FOXF2 | |
| FOXF2 (NP_001443, 346 a.a. ∼ 443 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Western Blot | |
| 2F5 | |
| Mouse monoclonal antibody raised against a partial recombinant FOXF2. | |
| FOXF2 | |
| FKHL6/FREAC2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2295 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.