missing translation for 'onlineSavingsMsg'
Learn More
Learn More
formin 2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FMN2.
Brand: Abnova H00056776-A01.50uL
This item is not returnable.
View return policy
Description
Formin homology (FH) domain proteins (e.g., FMN; MIM 136535) play a role in cytoskeletal organization and/or establishment of cell polarity.[supplied by OMIM
Sequence: DQEAEENSLTETHKCFLETTAYFFMKPKLGEKEVSPNAFFSIWHEFSSDFKDFWKKENKLLLQERVKEAEEVCRQKKGKSLYKIKPRHDSGIKAKISMKTSpecifications
| formin 2 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant FMN2. | |
| FMN2 | |
| FMN2 | |
| FMN2 (AAH14364.2, 144 a.a. ∼ 243 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| Yes | |
| 56776 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC014364 | |
| Mouse | |
| 50 μL | |
| Signal Transduction | |
| Primary | |
| Human | |
| Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction