missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fluorescein/Oregon Green Rabbit anti-Chemical, Alexa Fluor™ 594, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen A11091
This item is not returnable.
View return policy
Description
Anti-fluorescent dye antibodies recognize specific fluorophores and, in most cases, quench their fluorescence. Thus many anti-dye antibodies, including those that recognize fluorescein, can serve as cell-impermeant probes for determining whether fluorescent dye-conjugated ligands, proteins, bacteria or other biomolecules have been internalized by endocytic or pinocytic processes.
Specifications
| Fluorescein/Oregon Green | |
| Polyclonal | |
| Alexa Fluor 594 | |
| FITC | |
| Fluorescein/Oregon Green | |
| 500 μL | |
| Primary | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunohistochemistry | |
| 1 mg/mL | |
| 0.1M potassium phosphate with 5mM sodium azide; pH 8 | |
| Rabbit | |
| Purified | |
| RUO | |
| 4° C, store in dark | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction