missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FLI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Description
PA5-16899 targets Fli-1 in WB applications and shows reactivity with Human, mouse, and Rat samples. The PA5-16899 immunogen is synthetic peptide derived from C-terminus of human Fli-1.
Ets-1 is the prototype member of a family of genes identified on the basis of homology to the v-Ets oncogene isolated from the E26 erythroblastosis virus. This family of genes currently includes Ets-1, Ets-2, Erg-1, Erg-2, Elk, E74, Fli-1, PU.1 and PEA3. Members of the Ets gene family exhibit varied pattern of tissue expression, and share a highly conserved carboxy terminal domain containing a sequence related to the SV40 large T antigen nuclear localizartion signal sequence.
Specifications
Specifications
| Antigen | FLI1 |
| Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.2% BSA and 15mM sodium azide; pH 7.6 |
| Gene | FLI1 |
| Gene Accession No. | Q01543, P26323 |
| Gene Alias | Friend leukemia integration 1 transcription factor, proto-oncogene Fli-1, transcription factor ERGB |
| Gene Symbols | FLI1 |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?