Learn More
Invitrogen™ FKBP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA1020
Description
PA1-020 detects FK506 binding protein 51 kDa (FKBP51) in canine, hamster, human, mouse and rat tissues. PA1-020 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~51 kDa protein representing FKBP51 in HeLa cell lysate. By Western blot PA1-020 also detects FKBP51 in whole tissue extracts from rat kidney and rat and mouse testes. The PA1-020 immunogen is a synthetic peptide corresponding to the residues C(394) Q K K A K E H N E R D R R(407) of human FKBP51. There is a single amino acid substitution (K396 to R396) in this domain between human and mouse FKBP51 protein. PA1-020 immunizing peptide (Cat. # PEP-078) is available for use in neutralization and control experiments.
FKBP5 is a member of the immunophilin protein family. This family plays a role in immunoregulation and basic cellular processes involving protein folding and trafficking. FKBP5 is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. The gene that encodes FKBP5 has been found to have multiple polyadenylation sites. Alternative splicing results in multiple transcript variants.
Specifications
| FKBP5 | |
| Polyclonal | |
| Unconjugated | |
| FKBP5 | |
| 51 kDa FK506-binding protein; 51 kDa FKBP; 54 kDa progesterone receptor-associated immunophilin; AIG6; androgen-regulated protein 6; D17Ertd592e; Dit1; EC 5.2.1.8; FF1 antigen; FK506 binding protein 5; FK506-binding protein 5; FKBP prolyl isomerase 5; Fkbp5; FKBP-5; FKBP51; FKBP-51; FKBP54; HSP90-binding immunophilin; p54; peptidylprolyl cis-trans isomerase; Peptidyl-prolyl cis-trans isomerase FKBP5; PPIase; PPIase FKBP5; Ptg-10; rotamase; T-cell FK506-binding protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100760904, 14229, 2289, 361810, 481759 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1mg/mL BSA and 0.05% sodium azide | |
| Q13451, Q64378 | |
| FKBP5 | |
| Synthetic peptide corresponding to the residues C(394) Q K K A K E H N E R D R R(407) of human FKBP51. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Hamster | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.