missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium Sensing Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA533384
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Senses changes in the extracellular concentration of calcium ions. The activity of this receptor is mediated by a G- protein that activates a phosphatidylinositol-calcium second messenger system.Specifications
Calcium Sensing Receptor | |
Polyclonal | |
Unconjugated | |
CASR | |
BoPCaR1; Ca sensing receptor; Ca(2+)-sensing receptor; calcium sensing receptor; calcium-sensing receptor; Calcium-sensing receptor (hypocalciuric hypercalcemia 1 severe neonatal hyperparathyroidism); Calcium-sensing receptor (hypocalciuric hypercalcemia 1, severe neonatal hyperparathyroidism); CaR; CaSR; CaSR antibody; cation sensing receptor; EIG8; Extracellular calcium-sensing receptor; FHH; FIH; G protein coupled receptor, family C, group 2, member A; G protein-coupled receptor, family C, group 2, member A; Gprc2a; HHC; HHC1; HYPOC1; LOW QUALITY PROTEIN: extracellular calcium-sensing receptor; NSHPT; parathyroid Ca(2+)-sensing receptor 1; parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1; PCaR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100034084, 100341543, 100520980, 100766791, 12374, 281038, 418266, 488007, 846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
O62714, P35384, P41180, Q9QY96 | |
CASR | |
Synthetic 17 amino acid peptide from N-terminal extracellular domain of human CASR. | |
50 μg | |
Primary | |
Bovine, Canine, Chicken, Equine, Hamster, Human, Mouse, Non-human Primate, Porcine, Rabbit | |
Antibody | |
IgG |