Learn More
anti-Fibronectin, Polyclonal, Abcam™
This product is for research use only and not for diagnostic or therapeutic procedures. Customer is granted a limited, non-exclusive license for internal research and shall not attempt to reverse engineer.
Brand: Abcam AB23750-100UG
Description
Host Species: Rabbit
Isotype: IgG
Cross Reactivity: Mouse, Human
Immunogen: Purified fibronectin from murine plasma
Applications:
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blotting
Specifications
| Fibronectin | |
| 1mg/mL | |
| Rabbit | |
| 100 μg | |
| Primary | |
| IgG |
| Polyclonal | |
| Unconjugated | |
| Purified fibronectin from murine plasma | |
| Cancer and Tumor Biology, Cardiovascular, Developmental Biology, Stem Cell Biology | |
| Human, Mouse |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.