Learn More
fragile histidine triad gene, Mouse, Clone: 1E3, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FHIT.
Brand: Abnova H00002272-M03.100ug
Description
This gene, a member of the histidine triad gene family, encodes a diadenosine 5',5‴-P1,P3-triphosphate hydrolase involved in purine metabolism. The gene encompasses the common fragile site FRA3B on chromosome 3, where carcinogen-induced damage can lead to translocations and aberrant transcripts of this gene. In fact, aberrant transcripts from this gene have been found in about half of all esophageal, stomach, and colon carcinomas. [provided by RefSeq]
Sequence: VVPGHVLVCPLRPVERFHDLRPDEVADLFQTTQRVGTVVEKHFHGTSLTFSMQDGPEAGQTVKHVHVHVLPRKAGDFHRNDSIYEELQKHDKEDFPASWRSpecifications
| fragile histidine triad gene | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| BC032336 | |
| FHIT | |
| FHIT (AAH32336, 31 a.a. ∼ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Endocrine/Metabolism | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1E3 | |
| Mouse monoclonal antibody raised against a partial recombinant FHIT. | |
| FHIT | |
| AP3Aase/FRA3B | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 2272 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.