Learn More
FHIT, Mouse, Clone: 1A2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FHIT.
Brand: Abnova H00002272-M02A.200uL
Description
This gene, a member of the histidine triad gene family, encodes a diadenosine 5',5‴-P1,P3-triphosphate hydrolase involved in purine metabolism. The gene encompasses the common fragile site FRA3B on chromosome 3, where carcinogen-induced damage can lead to translocations and aberrant transcripts of this gene. In fact, aberrant transcripts from this gene have been found in about half of all esophageal, stomach, and colon carcinomas. [provided by RefSeq]
Sequence: VVPGHVLVCPLRPVERFHDLRPDEVADLFQTTQRVGTVVEKHFHGTSLTFSMQDGPEAGQTVKHVHVHVLPRKAGDFHRNDSIYEELQKHDKEDFPASWRSpecifications
| FHIT | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC032336 | |
| FHIT | |
| FHIT (AAH32336, 31 a.a. ∼ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 2272 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 1A2 | |
| Mouse monoclonal antibody raised against a partial recombinant FHIT. | |
| FHIT | |
| AP3Aase/FRA3B | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgM κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.